HIV TAT (47-57) Peptide
Supplier: Anaspec Inc
AS-60023-1
AS-60023-5
103005-824EA
252.31
CAD
103005-824
103005-826
HIV TAT (47-57) Peptide
Proteins and Peptides
This is the most characterized fragment of the HIV transactivator protein (TAT). This arginine-rich TAT peptide penetrates plasma membrane directly, but not through endocytosis.
Sequence: YGRKKRRQRRR
MW: 1559.9 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...