Human Ang I Peptide
Supplier: Anaspec Inc
AS-20627
AS-20628
102996-060EA
127.69
CAD
102996-060
102996-062
Human Ang I Peptide
Proteins and Peptides
This human Angiotensin I (Ang I) sequence also corresponds to horse, sheep, pig, and rat Ang I. Ang I is cleaved to Ang II by the angiotensin-converting enzyme (ACE). There is also evidence for non-angiotensin-converting enzyme-dependent conversion of Ang I to Ang II. Human chymase efficiently converts the 10-mer Ang I to the 8-mer hormone Ang II by splitting the Phe8-His9 bond in Ang I.
Sequence: DRVYIHPFHL
MW: 1296.5 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...