Human ACTH (1-24)
Supplier: Anaspec Inc
AS-20613
AS-20614
102998-424EA
178.46
CAD
102998-424
102998-426
Human ACTH (1-24)
Proteins and Peptides
Neuropeptide ACTH (adenocorticotropin) amino acids (1-39), Cat AS-20610, is the natural cleavage product from POMC (proopimelanocortin) processing; however, the first 24-amino acid sequence from the carboxyl end, ACTH (1-24) has full biological activity of the (1-39) peptide. Both ACTH (1-24) and (1-39) possess neurotropic activity.
Sequence: SYSMEHFRWGKPVGKKRRPVKVYP
MW: 2933.5 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...