Human;Mouse;Rat Beta-Amyloid (25-35)
Supplier: Anaspec Inc
AS-24228
AS-24227-
102996-478EA
503.08
CAD
102996-478
103003-706
Human;Mouse;Rat Beta-Amyloid (25-35)
Proteins and Peptides
Aß (25-35) is the main factor responsible for Aß neurotoxic effects.
- Peptide Content: ≥ 60%