Human Beta-Amyloid (1-16)

Supplier: Anaspec Inc

AS-24225 AS-24226
103003-702EA 233.85 CAD
103003-702 103003-704
Human Beta-Amyloid (1-16)
Proteins and Peptides

Proteins interactions with reactive oxygen agents may result in covalent modifications of amino acid residues in proteins, formation of protein-protein cross-linkages and oxidation of the protein backbone resulting in protein fragmentation. Oxidation targets for Aß(1-16) are the histidine residues coordinated to the metal ions. Copper is bound to Aß in senile plaque of Alzheimer’s disease with Aß (1-16) taking part in the coordination of the Cu2+ ions. Cu2+ and Zn2+ are linked with the neurotoxicity of Aß and free radical damage.
Sequence: DAEFRHDSGYEVHHQK
Molecular Weight: 1955 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR