Human Beta-Amyloid (1-40), FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-23513-05
AS-23514-01
103003-540EA
1350.77
CAD
103003-540
103003-542
Human Beta-Amyloid (1-40), FAM (Carboxyfluorescein)
Proteins and Peptides
Fluorescent (FAM)-labeled ß-Amyloid (1-40), Abs/Em=494/521 nm. FAM is preferred over FITC because of its photo- and chemical stability.
- Peptide Content: ≥ 60%