Human [Pyr11]-beta-Amyloid (11-42)

Supplier: Anaspec Inc

AS-29903-01 AS-29903-1
103005-422EA 252.31 CAD
103005-422 103005-424
Human [Pyr11]-beta-Amyloid (11-42)
Proteins and Peptides

This is a pyroglutamic acid modified beta-amyloid 11-42 peptide. Pyroglutamic acid modified isoforms form the major isoforms with up to 20% of the total beta-amyloid species.
Sequence: Pyr-VHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
Molecular Weight: 3317.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR