Catalase Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
707002-480
707002-96
75817-340EA
1303.75
CAD
75817-340
75817-342
Catalase Assay Kit, Cayman Chemical Company
Assays
Enzyme Assays
For the measurement of catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.
- Assay 40 samples in duplicate
- Measure catalase activity down to 2 U/mL
- Plate-based colorimetric measurement (540 nm)
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilizes the peroxidatic function of catalase for determination of enzyme activity.