BD Trypticase™ Soy Broth Prepared Bottled Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
299113
299416
299416
CA-90005-184PK
662.8
CAD
CA-90005-184
90005-200
CA90005-200
BD Trypticase™ Soy Broth Prepared Bottled Media, BD Diagnostics
Media for Microbiology
Prepared Media for Microbiology
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples
- Use of highest quality BBL and Difco brand dehydrated culture media for preparation
- Conformation to current USP and EP requirements (see package labeling for details)
- Color-coded caps and/or labeling for most products, providing easy product identification
- Transparent labeling to allow a clear view of media and culture
Septum/Screw Cap style has a black, phenolic screw cap with septum.
They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.