BD BBL™ Fluid Thioglycollate Medium Prepared Bottled Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
257485
299417
299417
CA95062-834PK
289.33
CAD
CA95062-834
90005-202
CA90005-202
BD BBL™ Fluid Thioglycollate Medium Prepared Bottled Media, BD Diagnostics
Media for Microbiology
Prepared Media for Microbiology
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.
- Use of highest quality BBL and Difco brand dehydrated culture media for preparation
- Conformation to current USP and EP requirements (see package labeling for details)
- Color-coded caps and/or labeling for most products, providing easy product identification
- Transparent labeling to allow a clear view of media and culture
Septum/Screw Cap style has a black, phenolic screw cap with septum.
Wide mouth bottle holds 100 ml fill volume with lug cap.