Standard Methods Agar Prepared Bottled Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
299094 299102
CA90005-164PK 378.74 CAD
CA90005-164 CA90005-172
Standard Methods Agar Prepared Bottled Media, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.
  • Use of highest quality BBL and Difco brand dehydrated culture media for preparation
  • Conformation to current USP and EP requirements (see package labeling for details)
  • Color-coded caps and/or labeling for most products, providing easy product identification
  • Transparent labeling to allow a clear view of media and culture
Septum/Screw Cap style has a black, phenolic screw cap with septum.
Screw Cap is the same as Septum/Screw Cap but without septum.