Fluid A Prepared Bottled Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
290652 290821
CA90004-128PK 702.36 CAD
CA90004-128 CA90004-138
Fluid A Prepared Bottled Media, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.
  • Use of highest quality BBL and Difco brand dehydrated culture media for preparation
  • Conformation to current USP and EP requirements (see package labeling for details)
  • Color-coded caps and/or labeling for most products, providing easy product identification
  • Transparent labeling to allow a clear view of media and culture
Serum bottle holds up to 100 mL fill volume.
Septum/Screw Cap style has a black, phenolic screw cap with septum.