RODAC™ Contact Plates, TSA with Lecithin and Polysorbate 80, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
215277 215278
CA-90001-614PK 575.33 CAD
CA-90001-614 CA90004-226 CA90005-296 CA71003-766 CA71003-768 CA71003-770 CA71003-772 CA71003-786
RODAC™ Contact Plates, TSA with Lecithin and Polysorbate 80, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD RODAC™ contact plates are available with or without the BD RODAC™ Snap lid. The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage