RODAC™ Contact Plates with Snap Lid, TSA with Lecithin, Polysorbate 80, and Penicillinase, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
215273 215274
CA71003-762PK 1025.74 CAD
CA71003-762 CA71003-764
RODAC™ Contact Plates with Snap Lid, TSA with Lecithin, Polysorbate 80, and Penicillinase, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
Order Now
SPECIFICATIONS
Environmentally Preferable
Sterility Sterile