RODAC™ Contact Plates with Snap Lid, DE Neutralizing Agar, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
292646 292647 215280
CA90001-266PK 83.09 CAD
CA90001-266 CA90007-908 CA90007-910 CA71003-776
RODAC™ Contact Plates with Snap Lid, DE Neutralizing Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage