e-Myco PCR Detection Kits, Bulldog Bio

Supplier: Bulldog Bio

25233 2523348 25234 2523448
102407-866EA 816.92 CAD
102407-866 102407-868 102407-870 102407-872
e-Myco PCR Detection Kits, Bulldog Bio
Assays Nucleic Acid Assays

PCR Detection Kits greatly simplify testing and detection of Mycoplasma contamination in cell cultures


  • Detection method: Agarose Gel
  • Number of species detected: 49 (e-Myco); 209 (e-Myco Plus)
  • Target size: ~260 bp
  • Internal control: ~160 bp


It can identify the five most common cell culture-infecting species of Mycoplasma all the way down to 20 CFU/mL. It contain an internal control (primer set and template DNA) to identify false negatives caused by faulty PCR conditions and 8-methoxypsoralen (8-MOP) to prevent cross-contamination by PCR products from earlier experiments.


With PCR testing, reliable results are obtained within a few hours, since the presence of contaminant Mycoplasmas can be easily and sensitively detected by verifying the amplified DNA fragments after gel electrophoresis. These Mycoplasma PCR Detection Kits have been shown to be a highly sensitive, specific and rapid method for the detection of Mycoplasmas contamination in cell cultures.


Delivery information: PCR kits - 48 x 20 µL reactions in 8-strip lypholized tubes.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR