Anti-CASP8 Monoclonal Antibody [clone: C15]

Supplier: Adipogen
AG-20B-0057-C050 AG-20B-0057-C100
102971-596EA 400 CAD
102971-596 102971-598
Anti-CASP8 Monoclonal Antibody [clone: C15]
Antibodies
Procaspase-8 belongs to the family of caspases. Binding of FasL to Fas leads to formation of a receptor complex at the cellular membrane, which was named DISC. The DISC consists of oligomerized receptors, the DD-containing adaptor molecule FADD, procaspase-8, procaspase-10 and c-FLIP. The DISC structure provides a platform for the oligomerization of procaspase-8 that allows two procaspase-8 homodimers to be in the close proximity leading to the initial activation of procaspase-8. At the first cleavage step, the N-terminal p43/p41 and the C-terminal p30 cleavage products are generated. Importantly, these cleavage products already possess catalytic activity. At the second cleavage step, p43/p41 and p30 are processed to p10 and p18, respectively, which leads to the generation of the active caspase-8 heterotetramer (p18/p10)2.

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: C15
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR