Anti-DLK1 Monoclonal Antibody [clone: PF183E]

Supplier: Adipogen
AG-20A-0058Y-C050 AG-20A-0058Y-C100
102971-652EA 324.62 CAD
102971-652 102971-654
Anti-DLK1 Monoclonal Antibody [clone: PF183E]
Antibodies
The transmembrane and secreted protein delta-like 1 homolog (DLK1; Protein delta homolog 1; Preadipocyte factor 1; Pref-1) belongs to the EGF-like family. It can be cleaved by a tumor necrosis factor-alpha converting enzyme (TACE) to release a soluble form. DLK1 plays important roles in regulating cell differentiation such as adipogenesis and osteogenesis. Membrane-bound DLK1 promotes hypertrophic myotube formation, whereas soluble DLK1 inhibits myotube formation. The soluble form of DLK1 inhibits preadipocyte differentiation, while membrane DLK1 represses preadipocyte proliferation. DLK1 is also associated with tumor invasion.

Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: PF183E
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Rat IgG2akappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR