Potassium iodide ≥99.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

PO185-2.5KGP-C PO185-2.5KGPL
75812-054CS 10851.05 CAD
75812-054 75812-056
Potassium iodide ≥99.0% USP
Potassium iodide

Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Kl
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now

Specification Test Results

Assay 99.0 - 101.5 %
Alkalinity To Pass Test
Loss On Drying max. 1.0 %
Iodate max. 4 Ug/G
Nitrate, Nitrite, and Ammonia To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR