Chem-Wik® Rosin SD Desoldering Braids, ITW Chemtronics®
Supplier: ITW Chemtronics
10-5L 7-5L
33500-180EA 153.02 CAD
33500-180 33500-190 33500-200 33500-210
Chem-Wik® Rosin SD Desoldering Braids, ITW Chemtronics®
Soldering Supplies Desoldering Braids
Ultrapure, oxygen-free copper braids which quickly and completely remove solder from circuit boards and components.
  • Chem-Wik® Rosin packaged in static dissipative bobbins
  • Reduces the risk of damage associated with static electricity
  • Coated with ultra-high purity
These ultrapure, oxygen-free copper braids quickly and completely remove solder from circuit boards and components. The fast wicking action protects components from heat damage. Braids are coated with ultra-high purity, noncorrosive Type R rosin flux type. Meet DOD-STD-1686C, MIL-HDBK-263B.