BD BBL Buffered Sodium Chloride-Peptone Solution, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
257087 257086
CA95053-336PK 740.36 CAD
CA95053-336 CA95053-334
BD BBL Buffered Sodium Chloride-Peptone Solution, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Buffered sodium chloride pepton solution for test samples.
  • Dissolve, suspend and dilute test samples
  • TEar-off seal, stopper and pull-ring
  • 100 mL infusion bottle
Buffered sodium chloride peptone solution is used for dissolving, suspending and diluting test samples. 100 mL infusion bottle with flip tear-off seal, stopper and pull-ring.
Certifications: EP / USP compliant.