Difco™ 2XYT Medium, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
244010 244020
CA90003-328EA 474.74 CAD
CA90003-328 CA90003-330
Difco™ 2XYT Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
For growth and propagation of E
  • Dehydrated culture media
  • Sequencing and phage display research
  • Provides nitrogen and growth factor
A nutritionally rich growth medium designed for growth of recombinant strains of Escherichia coli. Also used for propagation of M13 bacteriophage for sequencing and phage display research. The components of Difco 2xYT Medium provide nitrogen and growth factors that allow bacteriophage to reproduce in large quantities without exhausting the host.
coli infected with single strand filamentous bacteriophage M13.