3B Scientific® Spinal Injection Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
1017891 1020629 1017892 1020800 1017893
470224-966EA 476.15 CAD
470224-966 470224-954 470231-522 470231-516 470231-518
3B Scientific® Spinal Injection Trainer
Models Human Biology Models
This epidural and spinal injection trainer is very impressive thanks to its extremely realistic haptic feedback, and the use of high-quality, hard-wearing materials.
  • Easy to clean
Thanks to close cooperation with epidural anesthesia specialists, incomparably realistic conditions could be recreated. The light, compact structure can be used in a wide variety of ways. The device has low operating costs and is intuitive to use, ensuring lasting, high-level performance.
A summary of the device's main functions:
Epidural anesthesia using the loss-of-resistance and the hanging-drop technique
Spinal anesthesia with realistic resistance of the dura and arachnoid mater with or without a cannula
Fluid-filled spinal canal with realistic outflow rate thanks to the ability to adjust the excess pressure
An epidural catheter can be inserted into the epidural space
Closed water system