Coventry™ Sealed Polyester Swabs, ITW Chemtronics®
Supplier: ITW Chemtronics
36060 38540 36061I 38040
15145-405CS 2329 CAD
15145-405 15145-409 83007-604 15145-569
Coventry™ Sealed Polyester Swabs, ITW Chemtronics®
Swabs Swabs for Cleaning
Swabs feature either knit or nonwoven fabric heads, and are acid and solvent resistant
  • High strength, absorbency, solvent compatibility, and absorbency rate
  • Does not generate loose fibers or particles
  • No adhesives or binders are used in the construction
  • Strongest seams available will not split, also good for tight tolerance work
  • Micro-fiber has increased number of microspace to trap more particles
Aqua-Prime™ process available on all sealed polyester swabs. Class 10 (ISO Class 4) processing gives low ionic, nonvolatile residue and particle contaminants.
Sealed edges ensure high strength with no loose fibers or particulates. They are ideal for tough scrubbing around raised shapes.