Difco™ HC Agar Base, BD
Supplier: BD CANADA MICROBIOLOGY CA
268510
268510
CA90003-802EA
202.24
CAD
CA90003-802
90003-802
Difco™ HC Agar Base, BD
Media for Microbiology
Prepared Media for Microbiology
For enumerating molds in cosmetics Use with Tween™ 80.