Difco™ Chocolate Agar Base (GC Medium), BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
228920 228930 228950
CA90002-012EA 879.9 CAD
CA90002-012 CA90002-014 CA90002-016
Difco™ Chocolate Agar Base (GC Medium), BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Use with Hemoglobin or Hemoglobin 2%, Antimicrobic Vial CNV or CNVT, Supplement A, B, C or VX
Cultivation of N. gonorrhoeae and other fastidious organisms.