Lysine Medium, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
211759 284920
CA90000-164EA 217.33 CAD
CA90000-164 CA90000-334 CA90004-052
Lysine Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Differential medium to detect organisms that decarboxylate or deaminate lysine and produce hydrogen sulfide.
Differentiation of bacteria based on ability to decarboxylate lysine.