BBL™ IsoVitaleX™ Enrichment Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
211875 211876
CA90000-412PK 285.7 CAD
CA90000-412 CA90000-414
BBL™ IsoVitaleX™ Enrichment Media, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Enrichment is a chemically defined supplement used as an additive to media.
  • Chemically-defined supplement
  • Cultivation of nutritionally fastidious microorganisms
  • Media additive
Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.