Difco™ Motility GI Medium, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
286910
286910
CA90004-070EA
452.33
CAD
CA90004-070
90004-070
Difco™ Motility GI Medium, BD Life Sciences
Media for Microbiology
Prepared Media for Microbiology
Motility GI Medium is used for detecting motility of microorganisms and for separating organisms in their motile phase.
- Beef heart infusion, peptone, and gelatin provide nitrogen and vitamins
- Agar is the solidifying agent
- Motility evidenced by diffuse growth away from line of inoculation
Nonmotile organisms grow only along the line of inoculation; the presence of diffuse growth away from the line or spot of inoculation is evidence of motility.