Microbial Content Test Agar, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
255310 255320
CA90003-498EA 1174.49 CAD
CA90003-498 CA90003-500
Microbial Content Test Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Microbial Content Test Agar is used for the detection of microorganisms on surfaces sanitized with quaternary ammonium compounds.
  • Casein and peptones provide essential nutrients
  • Sodium chloride maintains osmotic equilibrium
  • Agar is the solidifying agent
Microbial Content Test Agar is also known as Tryptic Soy Agar with Lecithin and Polysorbate 80, and is recommended for the Aerobic Plate Count (Microbial Limit Test) for water-miscible cosmetic products containing preservatives.