Difco™ Malt Agar, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
224100 224200
CA90001-762EA 9489.06 CAD
CA90001-762 CA90001-772
Difco™ Malt Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Malt Agar is used for isolating and cultivating yeasts and molds from food and for cultivating yeast and mold stock cultures.
  • Malt extract provides carbon, protein, and essential nutrients
  • Acidic pH level
  • Agar is the solidifying agent
The acidic pH of Malt Agar allows for optimal growth of molds and yeasts while restricting bacterial growth.