3B Scientific® SONOtrain™ Ultrasound Breast Phantoms
Supplier: American 3B Scientific
1019634 1019635 1019649 1019650 1024722 1024723
470223-776EA 1296.65 CAD
470223-776 470223-778 470224-944 470224-946 470359-124 470359-126
3B Scientific® SONOtrain™ Ultrasound Breast Phantoms
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
The material of the SONOtrain trainer simulates the feel of real soft tissue to carry out palpations and injections and shows realistic texture and echogenicity on ultrasound images.
  • Easy to Use
  • Cost Effective
  • Realistic Echogenicity
  • Available with Cysts or Tumors
  • For Basic Training and Improving Dexterity
The SONOtrain trainer is an excellent teaching tool for basic training and for improving dexterity and hand-eye coordination skills.
The Phantom with cysts includes two cysts for practicing punctures and aspirations, and the Phantom with tumors contains three tumors for training on punctures or needle biopsy.
Ordering information: An ultrasound device is not part of the delivery content.