3B Scientific® SONOtrain™ Ultrasound Foreign Body Phantom
Supplier: American 3B Scientific
1019636 1019651 1024720
470223-772EA 1692.95 CAD
470223-772 470224-948 470359-132
3B Scientific® SONOtrain™ Ultrasound Foreign Body Phantom
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Real soft tissue feel.
  • Easy to Use
  • Cost Effective
  • Realistic Echogenicity
  • Trainer Simulates the Feel of Real Soft Tissue
  • Six Different Foreign Bodies Provided
The material of the SONOtrain trainer simulates the feel of real soft tissue to carry out palpations and injections and shows realistic texture and echogenicity on ultrasound images. The SONOtrain trainer is an excellent teaching tool for basic training and for improving dexterity and hand-eye coordination skills.
The ultrasound block comes with six different foreign bodies for insertion. Pellets, shard of glass, nail, projectile, loose chippings, and splinters of wood can be clearly detected and recognized.
Ordering information: An ultrasound device is not part of the delivery content.