Mycosel™ Agar Slants, BD
Supplier: BD CANADA MICROBIOLOGY CA
220967 211462
CA-90001-168PK 538.17 CAD
CA-90001-168 CA90000-202 CA90001-166
Mycosel™ Agar Slants, BD
Media for Microbiology Prepared Media for Microbiology
Mycosel™ Agar is a selective medium for the isolation of pathogenic fungi from materials having a mixed flora of other fungi and bacteria.
  • Soybean meal peptones provide nutrients
  • Dextrose is an energy source
  • Cycloheximide inhibits most saprophytic molds
  • Chloramphenicol inhibits most gram-positive and gram-negative bacteria
Mycosel™ Agar was developed by using the ingredients of Mycophil™ Agar as a nutritive base to which cycloheximide and chloramphenicol were added as selective agents. It is widely used for the isolation of fungi from a variety of sources, and is recommended for the recovery of dermatophytes.