BBL™ Lowenstein-Jensen Medium Slants, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
220909 220908
CA90001-156CS 808.65 CAD
CA90001-156 CA90001-154
BBL™ Lowenstein-Jensen Medium Slants, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
BBL™ L-J Medium is a selective medium for the isolation of mycobacteria from specimens containing mixed flora.
  • Contains inorganic salts and other sources of nitrogen
  • Glycerol provides source of carbon and energy
  • Egg yolk is a source of lipids
  • Malachite green dye partially inhibits growth of other bacteria
BD BBL™ Mycobactosel™ L-J Medium is Lowenstein-Jensen Medium1-3 plus cycloheximide, lincomycin and nalidixic acid for use with specimens likely to contain many contaminating organisms.