EDTA Buffers 0.5 M, Lonza
Supplier: LONZA WALKERSVILLE INC. CA
51201 51234
CA12001-382EA 79.13 CAD
CA12001-382 CA12001-410
EDTA Buffers 0.5 M, Lonza
Buffers Cloning Buffers
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
EDTA (ethylenediaminetetraacetic acid) is a polyamino carboxylic acid. When in water, It is a useful chelating agent due to its ability to sequester metal ions. EDTA solutions are able to limit, reduce or eliminate various undesirable effects in molecular biological and cell culture processes due to the presence of these metal. After being bound by EDTA, these metal ions remain in the solution but exhibit diminished reactivity. The use of EDTA solutions allow for greater experimental control.
Store all reagents at 18 to 26 °C (64 to 79 °F).
Each reagent is prepared with 18 megOhm water, filtered using a 0.2 μm filter, and filled into sterile bottles.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2942bfbab5e0186802dddb9e3245c63386620a75&vid=4eed438c0bb148e5671fafe1570c1d816c0bf477&prid=web&clid=SAPDAM