Formalin 20% Fixative, neutral buffered (20% NBF)

Supplier: AZER SCIENTIFIC CA
Danger

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

20NBF-4-G 20CUNBF5G
CA75788-508CS 548.59 CAD
CA75788-508 CA75788-510
Formalin 20% Fixative, neutral buffered (20% NBF)
Formalin

General purpose fixative for surgical and necropsy specimens.


  • Phosphate buffered to neutral pH
  • 7-8% percent formaldehyde concentration


With a higher concentration of formaldehyde, this solution will achieve complete and rapid fixation in even the most dense tissues.


Packaging: 5 gallon cube is equipped with a dispensing spigot.

Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR