BBL™ Thioglycollate Medium, Enriched with Vitamin K1 and Hemin, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
221742 221787
CA-90001-508PK 215.33 CAD
CA-90001-508 CA90001-520
BBL™ Thioglycollate Medium, Enriched with Vitamin K1 and Hemin, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Thioglycolate Medium, Enriched with Vitamin K1 and Hemin is a general-purpose medium for the cultivation of fastidious and nonfastidious microorganisms.
  • Digests of casein and soybean meal provide essential nutrients
  • Dextrose is a source of energy
  • Sodium chloride maintains osmotic equilibrium
Enriched broth medium is recommended for use in the isolation and cultivation of fastidious or slow-growing, obligately anaerobic microorganisms present in clinical materials.