BBL™ Saline, Normal, BD
Supplier: BD CANADA MICROBIOLOGY CA
221819 297815
CA-90001-538PK 264.53 CAD
CA-90001-538 CA-90005-030
BBL™ Saline, Normal, BD
Media for Microbiology Prepared Media for Microbiology
Normal saline is used in procedures that require the use of an isotonic diluent.
  • Ready to use
  • Provide ensured cell integrity
  • Used with a vast array of procedures
An isotonic diluent may be used for dilution of bacterial cells to provide a concentration suitable for microscopic observation, determination of cell numbers, analysis for genetic or metabolic properties, washing cells preparatory to study, or preparation of standardized inocula.