BBL™ Sabouraud Dextrose Agar (Emmons), Slants, BD
Supplier: BD CANADA MICROBIOLOGY CA
221013
221825
CA-90001-186PK
534.75
CAD
CA-90001-186
CA-90001-544
CA90001-184
CA90006-488
BBL™ Sabouraud Dextrose Agar (Emmons), Slants, BD
Media for Microbiology
Prepared Media for Microbiology
Sabouraud Dextrose Agar, Emmons is a nonselective medium for the cultivation and maintenance of fungi.
- Peptones provide a source of nitrogenous growth factors
- Dextrose provides an energy source
- Agar is the solidifying agent
Sabouraud Dextrose Agar, Emmons is a peptone medium supplemented with dextrose to support the growth of fungi.