BBL™ OF Medium, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
221328 221705 221703 221714
CA90001-380PK 67.37 CAD
CA90001-380 CA90001-308 CA90001-486 CA90001-484 CA90001-492
BBL™ OF Medium, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
OF Basal Medium, when supplemented with an appropriate carbohydrate, is used to determine the oxidative and fermentative metabolic activities of gram-negative bacilli.
  • Carbohydrate provides a source of energy
  • Dipotassium phosphate adds buffering capacity to the medium
  • Agar permits determination of motility
Changed in the covered agar are considered to be due to true fermentation, while changes in the open tubes are due to oxidative utilization of the carbohydrate present. If the carbohydrate is not utilized by either method, there is no acid production in either tube.