Tryptic Soy Broth (TSB) with Lecithin and Tween®
Supplier: HARDY DIAGNOSTICS
U306 U241 U261 U292 K134 U134G U423 U425 U229
10053-678EA 435.04 CAD
10053-678 10124-508 CA89408-114 CA89408-116 CA89429-336 CA10127-494 CA76047-198 CA10124-506 CA10053-676 CA89408-126
Tryptic Soy Broth (TSB) with Lecithin and Tween®
Media for Microbiology Dehydrated Media for Microbiology
Recommended for use in the cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • For fastidious and nonfastidious microorganisms
  • With neutralizing additives
Lecithin and Tween® 80 help in neutralizing residual sanitizing chemicals and help in emulsifying samples. This product is not intended for use in diagnosing human disease.
Ordering information: This product is not intended to be used for the diagnosis of human disease.
Order Now
SPECIFICATIONS
Environmentally Preferable