Hygiena SuperSnap™, High-Sensitivity ATP surface test, Hardy Diagnostics
Supplier: Hardy Diagnostics
SUS3000 SUS3000
89492-624EA 732.99 CAD
89492-624 CA89492-624
Hygiena SuperSnap™, High-Sensitivity ATP surface test, Hardy Diagnostics
Monitoring Systems Hygiene Monitoring Systems
SuperSnap™ is the most sensitive ATP surface test in Hygiena's ATP hygiene monitoring portfolio of products. SuperSnap™ is an all-in-one test which detects extremely low levels of ATP (Adenosine Triphosphate).
  • Sensitivity - Up to 100x better than other systems
  • Allergen prevention
  • Process monitoring for perishable/short-life foods and beverages
  • Industrial applications with strict cleaning protocols
  • Improved detection at very low ATP values (e.g. dairies and beverage processors)
  • Improved Buffer Capacity
  • Extremely resistant to harsh or concentrated chemical
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable