Saline, 0.85%
Supplier: HARDY DIAGNOSTICS
K52 U157 U157 K248 R47
10052-548EA 105.43 CAD
10052-548 10142-322 CA10142-322 CA76168-994 76411-522
Saline, 0.85%
Media for Microbiology Additives and Supplements for Microbiology
For the preparation of isotonic suspensions and dilutions of microorganisms.
Saline products are recommended for procedures that require the use of an isotonic diluent. It is also recommended foruse in preparing test suspensions of organisms.