Neutralizing Buffered Peptone Water
Supplier: HARDY DIAGNOSTICS
U181 K281 C9243 C9240 C9241 C9242
75813-986PK 428.04 CAD
75813-986 75838-644 75838-646 CA75873-672 CA76006-298 CA76006-300 CA76006-302
Neutralizing Buffered Peptone Water
Media for Microbiology Dehydrated Media for Microbiology
Used in the recovery of sub-lethally injured Salmonella species from industrial samples prior to selective enrichment and isolation.
This medium is recommended by the USDA FSIS (United States Department of Agriculture, Food Safety and Inspection Service) for poultry verification sampling.