Middlebrook 7H9 Broth
Supplier: HARDY DIAGNOSTICS
C62 C32
10127-468EA 214.37 CAD
10127-468 CA76006-348
Middlebrook 7H9 Broth
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of Mycobacterium spp. from sterile body sites.
Hardy Diagnostics Middlebrook 7H9 Broth and Middlebrook 7H9 Broth with Tween® 80 are recommended for use in the cultivation of Mycobacterium spp. from sterile fluids. The media are also used for preparing dilutions of mycobacteria for antimicrobial testing.