Czapek-Dox and Czapek Solution Media, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
233810 233910 233910
CA90002-654EA 162.3 CAD
CA90002-654 CA90002-658 90002-658
Czapek-Dox and Czapek Solution Media, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Czapek-Dox Broth and Czapek Solution Agar are used for cultivating fungi and bacteria capable of using inorganic nitrogen.
  • Saccharose is the sole carbon source
  • Sodium nitrate is the sole nitrogen source
  • Dipotassium phosphate is the buffering agent
  • Potassium chloride contains essential ions
  • Magnesium sulfate and ferrous sulfate are sources of cations
  • Agar is the solidifying agent in Czapek Solution Agar
Czapek-Dox media are useful in a variety of microbiological procedures, including soil microbiology and fungi and mildew resistance tests. These media produce moderately vigorous growth of most saprophytic aspergilli and yield characteristic mycelia and conidia.