Difco™ Columbia Blood Agar, BD
Supplier: BD CANADA MICROBIOLOGY CA
Columbia Blood Agar is an infusion-free basal medium to use with or without blood for the cultivation of fastidious microorganisms.
- Used with blood in isolating and cultivating fastidious microorganisms
- Used in qualitative procedures
- For isolating and cultivating Haemophilus species from clinical specimens
Columbia Blood Agar is utilized as the base for media containing blood and for selective media formulations in which various combinations of antimicrobial agents are used as additives.
Columbia Blood Agar Base EH (Enhanced Hydrolysis) is used with blood in isolating and cultivating fastidious microorganisms.