BBL™ Columbia Agar, BD
Supplier: BD CANADA MICROBIOLOGY CA
211126 211124 279240 279240
CA90000-096EA 4069.64 CAD
CA90000-096 CA90000-092 CA90004-012 90004-012
BBL™ Columbia Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Columbia Agar Base is a highly nutritious, general-purpose medium for the isolation and cultivation of nonfastidious and fastidious microorganisms from a variety of clinical materials.
  • Used with blood in isolating and cultivating fastidious microorganisms
  • Used in qualitative procedures
  • For isolating and cultivating Haemophilus species from clinical specimens
Columbia Agar Base is utilized as the base for media containing blood and for selective media formulations in which various combinations of antimicrobial agents are used as additives.