Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
237200
237500
CA90003-032EA
1459.31
CAD
CA90003-032
CA90003-038
CA90003-040
Bacto™ Brain Heart Infusion, BD Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Brain Heart Infusion (BHI) Broth is a general-purpose liquid medium for the growth of a wide variety of bacterial and fungal species.
- Nutrients come from peptones and infusions of brain and heart
- Supports the growth of fastidious and nonfastidious microorganisms
BHI Broth is used for the cultivation of a wide variety of microorganisms, including bacteria, yeasts, and molds.